missing translation for 'onlineSavingsMsg'
Learn More

Diethyl iminodiacetate, 97%

Product Code. 15447088
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447088 5 mL 5mL
15457088 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447088 Supplier Thermo Scientific Alfa Aesar Supplier No. H60608.AA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6290-05-7 | C8H15NO4 | 189.21 g/mol

It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is mainly used for the manufacture of N-(Phosphonomethyl) iminodiacetic acid and glyphosate. Used as raw material for ion exchange resin; for rubbers, electroplating and food additives.

Solubility
Moderately soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6290-05-7
MDL Number MFCD00041925

Specifications

Density 1.056
Quantity 5 mL
Refractive Index 1.435
Molecular Formula C8H10ClN
Linear Formula HN(CH2CO2C2H5)2
Synonym 1-4-chlorophenyl ethanamine, 1-4-chlorophenyl ethylamine, 4-chloro-alpha-methylbenzylamine, 1-4-chlorophenyl ethan-1-amine, 4-chloro-alpha-methyl benzylamine, 1-4-chlorophenyl-1-ethanamine, 1-4-chloro-phenyl-ethylamine, acmc-20amkz, acmc-20apje, acmc-209jma
Boiling Point 208°C
Solubility Information Moderately soluble in water.
InChI Key PINPOEWMCLFRRB-UHFFFAOYNA-N
SMILES CC(C1=CC=C(C=C1)Cl)N
IUPAC Name 1-(4-chlorophenyl)ethanamine
Molecular Weight (g/mol) 155.63
PubChem CID 97803
Formula Weight 189.21
Percent Purity 97%
Chemical Name or Material Diethyl iminodiacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.