missing translation for 'onlineSavingsMsg'
Learn More

Diethyl iminodiacetate, 98%

Product Code. 10647371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647371 100 g Glass bottle 100g
10196903 5 g Glass bottle 5g
10517552 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10647371 Supplier Thermo Scientific Acros Supplier No. 407321000
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6290-05-7 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6290-05-7
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00041925
InChI Key LJDNMOCAQVXVKY-UHFFFAOYSA-N
Synonym diethyl iminodiacetate, diethyl 2,2'-azanediyldiacetate, diethyliminodiacetate, iminodiacetic acid diethyl ester, glycine, n-2-ethoxy-2-oxoethyl-, ethyl ester, diethyl 2,2'-iminodiacetate, n,n-bis ethoxycarbonylmethyl amine, acetic acid, iminodi-, diethyl ester, ethyl 2-2-ethoxy-2-oxoethyl amino acetate, zlchem 799
PubChem CID 80502
IUPAC Name ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
SMILES CCOC(=O)CNCC(=O)OCC

Specifications

Color Yellow
Density 1.0800g/mL
Boiling Point 248.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HN(CH2COOC2H5)2
Refractive Index 1.4350 to 1.4370
Quantity 100 g
Specific Gravity 1.08
Formula Weight 189.21
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material Diethyl iminodiacetate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl iminodiacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.