missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Iminodiacetate, TRC

Product Code. 30377265
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377265 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377265 Supplier Toronto Research Chemicals Supplier No. TRCD4422702.5G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Iminodiacetate
CAS 6290-05-7
Molecular Formula C8H15NO4
Quantity 2.5 g
Synonym Iminodiacetic Acid Diethyl Ester, Iminodiethyl Diacetate, N,N-Bis(ethoxycarbonylmethyl)amine, N-(Ethoxycarbonylmethyl)glycine Ethyl Ester, NSC 6950, [[(Ethoxycarbonyl)methyl]amino]acetic Acid Ethyl Ester, 66: PN: US20050107325 PAGE: 155 claimed protein, Diethyl Iminobis(acetate)
InChI Formula InChI=1S/C8H15NO4/c1-3-12-7(10)5-9-6-8(11)13-4-2/h9H,3-6H2,1-2H3
SMILES CCOC(=O)CNCC(=O)OCC
IUPAC Name ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate
Molecular Weight (g/mol) 189.21
Formula Weight 189.1001
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.