missing translation for 'onlineSavingsMsg'
Learn More

Diethyl L-(+)-Tartrate, TRC

Product Code. 30372932
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372932 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30372932 Supplier Toronto Research Chemicals Supplier No. TRCD4450401G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (+)-Diethyl-L-Tartrate
CAS 87-91-2
Molecular Formula C8 H14 O6
Quantity 1 g
Synonym (2R,3R)-2,3-dihydroxy-butanedioic Acid 1,4-Diethyl Ester, (2R,3R)-2,3-dihydroxy-butanedioic Acid Diethyl Ester, , 2,3-Dihydroxy-[R-(R*,R*)]-butanedioic Acid Diethyl Ester, Diethyl Ester Tartaric Acid, (+)-(R,R)-Diethyl Tartrate, (+)-Diethyl L-Tartrate, (R,R)-Diethyl Tartrate, (R,R)-Tartaric Acid Diethyl Ester, Diethyl (+)-tartrate, Diethyl (2R,3R)-(+)-Tartrate, Diethyl (2R,3R)-2,3-Dihydroxysuccinate, Diethyl (2R,3R)-Tartrate, Diethyl (R,R)-(+)-Tartrate, Diethyl (R,R)-Tartrate, Diethyl 1,2-Dihydroxy-1,2-ethanedicarboxylate, Diethyl L-(+)-Tartrate, Diethyl L-(+)-Tartrate, Diethyl L-Tartarate, Diethyl L-Tartrate, Diethyl Tartrate, Ethyl (+)-Tartrate, Ethyl Tartrate, L-Diethyl Tartrate, L-Tartaric Acid Diethyl Ester, NSC 44808, NSC 8878, L-(+)-DET, (+)-DETA
InChI Formula InChI=1S/C8H14O6/c1-3-13-7(11)5(9)6(10)8(12)14-4-2/h5-6,9-10H,3-4H2,1-2H3/t5-,6-/m1/s1
SMILES CCOC(=O)[C@H](O)[C@@H](O)C(=O)OCC
IUPAC Name diethyl (2R,3R)-2,3-dihydroxybutanedioate
Molecular Weight (g/mol) 206.19
Formula Weight 206.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.