missing translation for 'onlineSavingsMsg'
Learn More

Diethyl malonate, 99+%

Product Code. 10070760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
25g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10070760 25 g Glass bottle 25g
10599240 500 g Glass bottle 500g
10192280 1 kg Glass bottle 1kg
10073943 2.5 kg Glass bottle 2.5kg
11935021 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10070760 Supplier Thermo Scientific Acros Supplier No. 114450250
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 105-53-3 | C7H12O4 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-53-3
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
InChI Key IYXGSMUGOJNHAZ-UHFFFAOYSA-N
Synonym diethyl malonate, malonic ester, propanedioic acid, diethyl ester, dicarbethoxymethane, malonic acid, diethyl ester, carbethoxyacetic ester, malonic acid diethyl ester, 1,3-diethyl propanedioate, ethyl propanedioate, ethyl malonate van
PubChem CID 7761
IUPAC Name diethyl propanedioate
SMILES CCOC(=O)CC(=O)OCC

Specifications

Melting Point -50.0°C
Density 1.0550g/mL
Boiling Point 199.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CCH2CO2C2H5
Refractive Index 1.4130 to 1.4150
Quantity 25 g
Beilstein 02,573
Fieser 10,182; 13,69
Merck Index 15,3878
Specific Gravity 1.055
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in ethanol, ether and acetone
Viscosity 2.14 mPa.s (10°C)
Formula Weight 160.17
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diethyl malonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.