missing translation for 'onlineSavingsMsg'
Learn More

Diethyl meso-2,5-dibromoadipate, 98%

Product Code. 11411250
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411250 25 g 25g
11401250 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411250 Supplier Thermo Scientific Alfa Aesar Supplier No. H26347.14
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 869-10-3 | C10H16Br2O4 | 360.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869-10-3
Molecular Formula C10H16Br2O4
Molecular Weight (g/mol) 360.04
MDL Number MFCD00075379
InChI Key UBCNJHBDCUBIPB-UHFFFAOYNA-N
Synonym diethyl 2,5-dibromoadipate, diethyl meso-2,5-dibromoadipate, 1,6-diethyl 2,5-dibromohexanedioate, 2,5-dibromoadipic acid diethyl ester, hexanedioic acid, 2,5-dibromo-, diethyl ester, diethyl2,5-dibromohexanedioate, 2,5-dibromohexanedioic acid diethyl ester, acmc-1bjoi, diethylmeso-dibromoadipate, diethylmeso-2,5-dibromoadipate
PubChem CID 241588
IUPAC Name diethyl 2,5-dibromohexanedioate
SMILES CCOC(=O)C(Br)CCC(Br)C(=O)OCC

Specifications

Melting Point 64°C to 67°C
Quantity 25 g
UN Number UN3261
Sensitivity Light sensitive
Formula Weight 360.04
Percent Purity 98%
Chemical Name or Material Diethyl meso-2,5-dibromoadipate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.