missing translation for 'onlineSavingsMsg'
Learn More

Diethyl methylmalonate, 99%

Product Code. 10615402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10615402 500 g Glass bottle 500g
10407750 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10615402 Supplier Thermo Scientific Acros Supplier No. 155145000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 609-08-5 | C8H14O4 | 174.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-08-5
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.2
MDL Number MFCD00009162
InChI Key UPQZOUHVTJNGFK-UHFFFAOYSA-N
Synonym diethyl methylmalonate, methylmalonic acid diethyl ester, diethyl 2-methylmalonate, propanedioic acid, methyl-, diethyl ester, 1,3-diethyl 2-methylpropanedioate, diethylmethylmalonate, ethyl methylmalonate, diethyl methylpropanedioate, malonic acid, methyl-, diethyl ester, 2-methylmalonic acid diethyl
PubChem CID 11857
IUPAC Name diethyl 2-methylpropanedioate
SMILES CCOC(=O)C(C)C(=O)OCC

Specifications

Color Colorless
Density 1.0100g/mL
Boiling Point 198°C to 199°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CCH(CH3)CO2C2H5
Refractive Index 1.4120 to 1.414
Quantity 500 g
Beilstein 02, 629
Fieser 05,216
Specific Gravity 1.01
Solubility Information Solubility in water: immiscible
Formula Weight 174.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl methylmalonate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.