missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Methylmalonate, TRC

Product Code. 30378067
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378067 5 g 5g
30379932 50 g -
30388131 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378067 Supplier Toronto Research Chemicals Supplier No. TRCD4444305G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-08-5
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.2
Synonym 2-Methylpropanedioic Acid 1,3-Diethyl Ester, Methylmalonic Acid Diethyl Ester, Diethyl 2-Methylmalonate, Diethyl Methylpropanedioate, Diethyl alpha-Methylmalonate, NSC 8700
SMILES CCOC(=O)C(C)C(=O)OCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H14O4/c1-4-11-7(9)6(3)8(10)12-5-2/h6H,4-5H2,1-3H3
Formula Weight 174.0892
Chemical Name or Material Diethyl Methylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.