missing translation for 'onlineSavingsMsg'
Learn More

Diethyl n-butylmalonate, 99%, Thermo Scientific™

Product Code. 11391937
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11391937 100 mL 100mL
11301947 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391937 Supplier Thermo Scientific Alfa Aesar Supplier No. L07539.AE
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-08-4
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00009170
InChI Key RPNFNBGRHCUORR-UHFFFAOYSA-N
Synonym diethyl butylmalonate, diethyl n-butylmalonate, butylmalonic acid diethyl ester, diethyl 2-butylmalonate, ethyl butylmalonate, propanedioic acid, butyl-, diethyl ester, unii-cyy2q744wb, cyy2q744wb, n-butylmalonic acid diethyl ester, malonic acid, butyl-, diethyl ester
PubChem CID 67244
IUPAC Name diethyl 2-butylpropanedioate
SMILES CCCCC(C(=O)OCC)C(=O)OCC

Specifications

Density 0.982
Boiling Point 236°C to 239°C
Flash Point 93°C (199°F)
Refractive Index 1.423
Quantity 100 mL
Beilstein 608785
Merck Index 14,1576
Formula Weight 216.28
Percent Purity 99%
Chemical Name or Material Diethyl n-butylmalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.