missing translation for 'onlineSavingsMsg'
Learn More

Diethyl n-dodecylphosphonate, ≥97%, Thermo Scientific Chemicals

Product Code. 15447807
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15447807 5 g 5g
15437807 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447807 Supplier Thermo Scientific Alfa Aesar Supplier No. H37697.06
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4844-38-6 | Molecular Formula: C16H35O3P | Molecular Weight: 306.427 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4844-38-6
Molecular Formula C16H35O3P
Molecular Weight (g/mol) 306.427
MDL Number MFCD00015595
InChI Key LRIWHHSVZCKNJL-UHFFFAOYSA-N
Synonym diethyl dodecylphosphonate, phosphonic acid, dodecyl-, diethyl ester, diethyl 1-dodecyl phosphonate, phosphonic acid, p-dodecyl-, diethyl ester, dodecylphosphonic acid diethyl ester, phosphonic acid,p-dodecyl-, diethyl ester
PubChem CID 78558
IUPAC Name 1-diethoxyphosphoryldodecane
SMILES CCCCCCCCCCCCP(=O)(OCC)OCC

Specifications

Color Colorless
Boiling Point 168°C (1.5 mmHg)
Quantity 5 g
Formula Weight 306.42
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Diethyl n-dodecylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.