missing translation for 'onlineSavingsMsg'
Learn More

Diethyl n-propylmalonate, 99%

Produktkod. 11480907
Change view
Klicka för att se tillgängliga alternativ
Quantity:
25 g
100 g
Förpackningsstorlek:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
11480907 100 g 100g
11470907 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11480907 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B23304.22
 Restricted Item
Estimated Shipment: 05-01-2027
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 2163-48-6 | C10H18O4 | 202.25 g/mol

Diethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).

Solubility
Soluble in Dichloromethane, Ethanol and Ethyl Acetate.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2163-48-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00009168
InChI Key GRRSDGHTSMJICM-UHFFFAOYSA-N
Synonym diethyl propylmalonate, diethyl 2-propylmalonate, 1,3-diethyl 2-propylpropanedioate, diethyl n-propylmalonate, diethylpropylmalonate, propylmalonic acid diethyl ester, propanedioic acid, propyl-, diethyl ester, diethyl-n-propylmalonate, malonic acid, propyl-, diethyl ester, diethyl propyl malonate
PubChem CID 16552
IUPAC Name diethyl 2-propylpropanedioate
SMILES CCCC(C(=O)OCC)C(=O)OCC

Specifikationer

Density 0.987
Boiling Point 221°C to 222°C
Flash Point 91°C (197°F)
Refractive Index 1.4205
Quantity 100 g
Solubility Information Soluble in Dichloromethane,Ethanol and Ethyl Acetate.
Formula Weight 202.25
Percent Purity 99%
Chemical Name or Material Diethyl n-propylmalonate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.