missing translation for 'onlineSavingsMsg'
Learn More

Diethyl oxalate, 99%

Product Code. 10487180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
25g
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10487180 25 g Glass bottle 25g
15700819 500 g Glass bottle 500g
10784651 1 kg Glass bottle 1kg
10347430 2.5 kg Glass bottle 2.5kg
10207263 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10487180 Supplier Thermo Scientific Acros Supplier No. 159180250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-92-1 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-92-1
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00009119
InChI Key WYACBZDAHNBPPB-UHFFFAOYSA-N
Synonym ethyl oxalate, diethyl ethanedioate, ethanedioic acid, diethyl ester, oxalic acid, diethyl ester, oxalic ether, diethyloxalate, ethyl oxalate van, oxalic acid diethyl ester, unii-860m3zwf6j, diethylester kyseliny stavelove
PubChem CID 7268
IUPAC Name diethyl oxalate
SMILES CCOC(=O)C(=O)OCC

Specifications

Acidity 0.01% max. (as oxalic acid)
Melting Point -41.0°C
Density 1.0700g/mL
Boiling Point 185.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CCO2C2H5
Refractive Index 1.4085 to 1.4105
Quantity 25 g
Beilstein 02, 535
Fieser 01,250; 02,132; 07,106; 15,133
Merck Index 15, 3142
Specific Gravity 1.07
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol,2-propanol and,acetone
Formula Weight 146.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl oxalate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl oxalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.