missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Oxalate, TRC

Product Code. 30375895
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30375895 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30375895 Supplier Toronto Research Chemicals Supplier No. TRCD4446755G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Oxalate
CAS 95-92-1
Molecular Formula C6H10O4
Quantity 5 g
Synonym Ethanedioic Acid 1,2-Diethyl Ester, Oxalic Acid Diethyl Ester, Diethyl Ethanedioate, Ethyl Oxalate, NSC 8851
InChI Formula InChI=1S/C6H10O4/c1-3-9-5(7)6(8)10-4-2/h3-4H2,1-2H3
SMILES CCOC(=O)C(=O)OCC
IUPAC Name diethyl oxalate
Molecular Weight (g/mol) 146.14
Formula Weight 146.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.