missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phenyl orthoformate, 97%

Product Code. 11437044
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11437044 50 g 50g
11427044 10 g 10g
11447044 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11437044 Supplier Thermo Scientific Alfa Aesar Supplier No. B21498.18
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14444-77-0 | C11H16O3 | 196.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14444-77-0
Molecular Formula C11H16O3
Molecular Weight (g/mol) 196.246
MDL Number MFCD00009228
InChI Key QTURWMMVIIBRRP-UHFFFAOYSA-N
Synonym diethyl phenyl orthoformate, diethoxymethoxy benzene, benzene, diethoxymethoxy, diethoxymethoxy-benzene, acmc-209csr, diethoxy phenoxy methane, diethoxymethoxy benzene #, diethylphenyl orthoformate, orthoformic acid diethyl phenyl ester
PubChem CID 84440
IUPAC Name diethoxymethoxybenzene
SMILES CCOC(OCC)OC1=CC=CC=C1

Specifications

Melting Point -26°C
Density 1.015
Boiling Point 103°C to 104°C (10 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.4795
Quantity 50 g
Beilstein 1872634
Sensitivity Moisture sensitive
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material Diethyl phenyl orthoformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.