missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phenylphosphonite, 98%, Thermo Scientific Chemicals

Product Code. 11426616
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426616 25 g 25g
11416616 5 g 5g
11436616 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426616 Supplier Thermo Scientific Alfa Aesar Supplier No. B24836.14
 Restricted Item
Estimated Shipment: 30-12-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 1638-86-4 | C10H15O2P | 198.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1638-86-4
Molecular Formula C10H15O2P
Molecular Weight (g/mol) 198.20
MDL Number MFCD00009086
InChI Key RVDJLKVICMLVJQ-UHFFFAOYSA-N
Synonym diethyl phenylphosphonite, diethoxyphenylphosphine, phosphonous acid, phenyl-, diethyl ester, diethyl benzenephosphonite, diethyl phenyl-phosphonite, phosphonous acid, p-phenyl-, diethyl ester, pubchem6466, diethylphenylphosphonite, diethyl phenylphophonite, acmc-20al8o
PubChem CID 74225
IUPAC Name diethoxy(phenyl)phosphane
SMILES CCOP(OCC)C1=CC=CC=C1

Specifikationer

Density 1.032
Boiling Point 64°C (0.7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.51
Quantity 25 g
Beilstein 2804900
Sensitivity Air sensitive
Formula Weight 198.2
Percent Purity 98%
Chemical Name or Material Diethyl phenylphosphonite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.