missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phosphate, 96%

Product Code. 15780869 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15780869 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15780869 Supplier Thermo Scientific Acros Supplier No. 407420050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-02-7 | C4H11O4P | 154.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 598-02-7, 7664-38-2
Melting Point 6.0°C
Density 1.2900g/mL
Boiling Point 203.0°C
Flash Point 91°C
Assay Percent Range 96%
Packaging Glass Bottle
Molecular Formula C4H11O4P
Linear Formula (C2H5O)2PO2H
Quantity 5 g
Specific Gravity 1.29
Synonym diethyl phosphate, diethyl acid phosphate, phosphoric acid, diethyl ester, diethyl phosphoric acid, diethylphosphate, diethylphosphoric acid, ethyl phosphate, di, o,o-diethyl hydrogen phosphate, ethyl phosphate, eto 2 ho po, o,o-diethylphosphoric acid
Solubility Information Solubility in water: slightly soluble.
InChI Key UCQFCFPECQILOL-UHFFFAOYSA-N
SMILES CCOP(=O)(O)OCC
IUPAC Name diethyl hydrogen phosphate
Molecular Weight (g/mol) 154.1
PubChem CID 654
ChEBI CHEBI:27708
Formula Weight 154.1
Percent Purity 96%
Chemical Name or Material Diethyl phosphate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.