missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phosphate-d10 (diethyl-d10), TRC

Product Code. 30409491
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409491 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30409491 Supplier Toronto Research Chemicals Supplier No. TRCP35943210MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Phosphate-d10 (Diethyl-d10)
CAS 2483831-48-5
Molecular Formula C4 D10 H O4 P
Quantity 10 mg
Synonym Ethan-1,1,2,2,2-d5-ol','1,1'-(hydrogen phosphate) (ACI), Ethan-1,1,2,2,2-d5-ol 1,1'-(hydrogen phosphate), Diethyl-d10 Phosphate, Diethyl Phosphate-d10, Diethyl Phosphate-D10
InChI Formula InChI=1S/C4H11O4P/c1-3-7-9(5,6)8-4-2/h3-4H2,1-2H3,(H,5,6)/i1D3,2D3,3D2,4D2
SMILES [2H]C([2H])([2H])C([2H])([2H])OP(=O)(O)OC([2H])([2H])C([2H])([2H])[2H]
IUPAC Name bis(1,1,2,2,2-pentadeuterioethyl) hydrogen phosphate
Molecular Weight (g/mol) 164.1631
Formula Weight 164.1023
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.