missing translation for 'onlineSavingsMsg'
Learn More

Diethyl (phthalimidomethyl)phosphonate, 97%

Product Code. 11444557
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444557 5 g 5g
11454557 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444557 Supplier Thermo Scientific Alfa Aesar Supplier No. B21096.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33512-26-4 | C13H16NO5P | 297.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33512-26-4
Molecular Formula C13H16NO5P
Molecular Weight (g/mol) 297.247
MDL Number MFCD00134313
InChI Key IUZMHUAHKBHJFY-UHFFFAOYSA-N
Synonym diethyl phthalimidomethyl phosphonate, diethyl 1,3-dioxoisoindolin-2-yl methyl phosphonate, phthalimidomethyl phosphonic acid diethyl ester, diethyl 1,3-dioxoisoindol-2-yl methylphosphonate, 2-diethoxycarbonyl methyl benzo c azoline-1,3-dione, acmc-1cp9b, diethyl phthalimidomethylphosphonate, n-diethoxyphosphoryl methyl phthalimide
PubChem CID 1924456
IUPAC Name 2-(diethoxyphosphorylmethyl)isoindole-1,3-dione
SMILES CCOP(=O)(CN1C(=O)C2=CC=CC=C2C1=O)OCC

Specifications

Melting Point 60°C to 64°C
Quantity 5 g
Formula Weight 297.25
Percent Purity 97%
Chemical Name or Material Diethyl (phthalimidomethyl)phosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.