missing translation for 'onlineSavingsMsg'
Learn More

Diethyl pyrocarbonate, 97%, BLD Pharm™

Product Code. 30783005
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30783005 10 g 10g
30782288 5 g 5g
30770289 25 g 25g
30771158 100 g 100g
30773092 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30783005 Supplier BLD Pharm Supplier No. BD12475810g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1609-47-8
Molecular Formula C6H10O5
MDL Number MFCD00009106
Quantity 10 g
SMILES O=C(OCC)OC(OCC)=O
Molecular Weight (g/mol) 162.14
Percent Purity 97%
Chemical Name or Material Diethyl pyrocarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.