missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Pyrocarbonate, MP Biomedicals™

Product Code. 11416672 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11416672 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11416672 Supplier MP Biomedicals Supplier No. 0215090280
  • ONLINE EXCLUSIVE PRICE
    505.00 €
    478.80 € / 100mL
    Save 26.20 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used to inactivate RNase enzymes. Diethyl Pyrocarbonate, MP Biomedicals is sensitive to moisture and pH. DEPC is also sensitive to ammonia, which causes decomposition to urethane, a possible carcinogen. It protects RNA against degradation and is effective

DEPC inactivates RNase in solution at about 0.1% (v/v). It inhibits the ryanodine binding to ryanodine/Ca2+ receptor channels in skeletal muscle in a dose- and time-dependent manner and increases the Ca2+ permeability of SR vesicles. DEPC is effective as a nuclease inhibitor with a variety of uses:

  • Modification reagent for His and Tyr residues in proteins
  • Robust probe for structural disruptions in dsDNA, reacting with fully or partially unstacked bases
  • Used to modify proteins and nucleic acids
  • PCR reaction for treating deionized water, which reduces the risk of RNA being degraded by RNases
  • Dot blot hybridization, to dilute total RNA isolated from different micro-organisms

Specifications

CAS 1609-47-8
Density 1.101g/mL at 25°C (Lit.)
Boiling Point 93°C to 94°C (18 mmHg)(literature)
Molecular Formula C6H10O5
Refractive Index n20/D 1.398 (Lit.)
MDL Number MFCD00009106
Quantity 100 mL
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
Solubility Information Soluble in alcohols, esters, ketones, hydrocarbons. A 50% w/w solution in ethanol may be prepared. Slowly soluble in water with hydrolytic decomposition yielding ethanol and CO2.
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
SMILES CCOC(=O)OC(=O)OCC
IUPAC Name ethoxycarbonyl ethyl carbonate
Molecular Weight (g/mol) 162.14
PubChem CID 3051
ChEBI CHEBI:59051
Formula Weight 162.10
Concentration or Composition (by Analyte or Components) Working Concentration: For Treatment of water to inhibit Rnase: Add 1mL DEPC to 1L water. Let sit overnight. Autoclave.
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl Pyrocarbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: diethyl pyrocarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.