missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Pyrocarbonate Ribonuclease inhibitor. MP Biomedicals

Product Code. 11406672 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11406672 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11406672 Supplier MP Biomedicals Supplier No. 0215090250
  • ONLINE EXCLUSIVE PRICE
    278.00 €
    263.55 € / 50mL
    Save 14.45 €
    5% Off
 Restricted Item
Estimated Shipment: 05-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Effective nuclease inhibitor used for modifying proteins and nucleic acids.

Specifications

CAS 1609-47-8
Density 1.12 g/mL
Molecular Formula C6H10O5
MDL Number MFCD00009106
Quantity 50 mL
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
SMILES CCOC(=O)OC(=O)OCC
IUPAC Name diethyl dicarbonate
Molecular Weight (g/mol) 162.14
PubChem CID 3051
ChEBI CHEBI:59051
Formula Weight 162.1
Physical Form Liquid
Chemical Name or Material Diethyl Pyrocarbonate, Ribonuclease inhibitor.
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl Pyrocarbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: diethyl pyrocarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.