missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Pyrocarbonate, TRC

Product Code. 30369290
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369290 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369290 Supplier Toronto Research Chemicals Supplier No. TRCD44204525G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1609-47-8
Molecular Formula C6H10O5
Quantity 25 g
Synonym Dicarbonic Acid, Diethyl Ester, Formic Acid, Oxydi-, Diethyl Ester, Pyrocarbonic Acid Diethyl Ester, Baycovin, DEPA, DEPC, Dekapex, Diethyl Dicarbonate, Diethyl Oxydiformate, Diethyl Pyrocarbonate, Piref, Ue 5908
InChI Formula InChI=1S/C6H10O5/c1-3-9-5(7)11-6(8)10-4-2/h3-4H2,1-2H3
SMILES CCOC(=O)OC(=O)OCC
Molecular Weight (g/mol) 162.14
Formula Weight 162.0528
Chemical Name or Material Diethyl Pyrocarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.