missing translation for 'onlineSavingsMsg'
Learn More

Diethyl sec-butylmalonate, 96%, Thermo Scientific™

Product Code. 11368705
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368705 25 g 25g
11358705 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368705 Supplier Thermo Scientific Alfa Aesar Supplier No. L12253.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-27-2
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.277
MDL Number MFCD00015194
InChI Key MIIZSUOEOUHAIZ-UHFFFAOYSA-N
Synonym diethyl sec-butylmalonate, diethyl 2-sec-butyl malonate, sec-butylmalonic acid diethyl ester, diethyl 1-methylpropyl malonate, diethyl 1-methylpropylmalonate, sec-butylmalonic acid, diethyl ester, 1,3-diethyl 2-sec-butyl propanedioate, diethyl s-butylmalonate, diethylsec-butylmalonate, acmc-209prt
PubChem CID 78940
IUPAC Name diethyl 2-butan-2-ylpropanedioate
SMILES CCC(C)C(C(=O)OCC)C(=O)OCC

Specifications

Density 0.98
Boiling Point 110°C to 114°C (18 mmHg)
Quantity 25 g
Beilstein 1709561
Formula Weight 216.28
Percent Purity 96%
Chemical Name or Material Diethyl sec-butylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.