missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Succinate, TRC

Product Code. 30380272
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380272 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380272 Supplier Toronto Research Chemicals Supplier No. TRCD4450601G

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Succinic acid diethyl ester
CAS 123-25-1
Molecular Formula C8 H14 O4
Quantity 1 g
Synonym Succinic acid-diethyl ester, Butanedioic acid, 1,4-diethyl ester (ACI), Butanedioic acid, diethyl ester (9CI), Succinic acid, diethyl ester (6CI, 8CI), 1,4-Diethyl butanedioate, Butandioic acid diethyl ester, Clorius, DESU, Diethyl 1,2-ethanedicarboxylate, Diethyl butanedioate, Diethyl succinate, Ethyl succinate, NSC 8875
InChI Formula InChI=1S/C8H14O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-6H2,1-2H3
SMILES CCOC(=O)CCC(=O)OCC
IUPAC Name diethyl butanedioate
Molecular Weight (g/mol) 174.19
Formula Weight 174.0892
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.