missing translation for 'onlineSavingsMsg'
Learn More

Diethyl sulfate, 98%

Product Code. 11319095
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319095 500 mL 500mL
11309095 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11319095 Supplier Thermo Scientific Alfa Aesar Supplier No. L14291.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 64-67-5 | C4H10O4S | 154.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-67-5
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00009099
InChI Key DENRZWYUOJLTMF-UHFFFAOYSA-N
Synonym sulfuric acid, diethyl ester, diethyl sulphate, diethylsulfate, diaethylsulfat, sulfuric acid diethyl ester, diethyl tetraoxosulfate, diaethylsulfat german, unii-k0fo4vfa7i, des van, diethylester kyseliny sirove
PubChem CID 6163
ChEBI CHEBI:34699
IUPAC Name diethyl sulfate
SMILES CCOS(=O)(=O)OCC

Specifications

Melting Point -25°C
Density 1.177
Boiling Point 208°C (decomposition)
Flash Point 104°C (219°F)
Refractive Index 1.4005
Quantity 500 mL
UN Number UN1594
Beilstein 1209714
Merck Index 14,3130
Formula Weight 154.19
Percent Purity 98%
Chemical Name or Material Diethyl sulfate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.