missing translation for 'onlineSavingsMsg'
Learn More

Diethyl sulfate, 99%

Product Code. 10560401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10560401 25 mL Glass bottle 25mL
10589410 1 L Glass bottle 1 L
10438450 2.5 L Glass bottle 2.5 L
10627763 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10560401 Supplier Thermo Scientific Acros Supplier No. 114560250
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 64-67-5 | C4H10O4S | 154.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-67-5
Molecular Formula C4H10O4S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00009099
InChI Key DENRZWYUOJLTMF-UHFFFAOYSA-N
Synonym sulfuric acid, diethyl ester, diethyl sulphate, diethylsulfate, diaethylsulfat, sulfuric acid diethyl ester, diethyl tetraoxosulfate, diaethylsulfat german, unii-k0fo4vfa7i, des van, diethylester kyseliny sirove
PubChem CID 6163
ChEBI CHEBI:34699
IUPAC Name diethyl sulfate
SMILES CCOS(=O)(=O)OCC

Specifications

Melting Point -24.0°C
Density 1.1795g/mL
Boiling Point 208.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2SO2
Refractive Index 1.3990 to 1.4010
Quantity 25 mL
Beilstein 01,327
Fieser 01,253
Merck Index 15,3149
Specific Gravity 1.1795
Solubility Information Solubility in water: 5g/L (20°C). Other solubilities: decomposes in water,soluble in alcohol and ether
Viscosity 1.79 mPa.s (20°C)
Formula Weight 154.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethyl sulphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Diethyl sulphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.