missing translation for 'onlineSavingsMsg'
Learn More

Diethyl sulfide, 96%

Product Code. 11322186
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322186 100 g 100g
11312186 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322186 Supplier Thermo Scientific Alfa Aesar Supplier No. L03493.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 352-93-2 | C4H10S | 90.18 g/mol

Diethyl sulfide is used as a solvent for anhydrous mineral salts and in plating baths for coating metals with gold or silver. It is used as a flavoring agent. It is also used in the addition reaction with platinum dichloride to prepare platinum ethynyl dimers and polymers with pendant ferrocenyl group. Further, it is used in the preparation of triethylsulfoniuim ethylsulfate by reaction with sulfuric acid diethyl ester. In addition to this, it is used in the quantification procedure of volatile sulfur compounds by solid-phase microextraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl sulfide is used as a solvent for anhydrous mineral salts and in plating baths for coating metals with gold or silver. It is used as a flavoring agent. It is also used in the addition reaction with platinum dichloride to prepare platinum ethynyl dimers and polymers with pendant ferrocenyl group. Further, it is used in the preparation of triethylsulfoniuim ethylsulfate by reaction with sulfuric acid diethyl ester. In addition to this, it is used in the quantification procedure of volatile sulfur compounds by solid-phase microextraction.

Solubility
Miscible with alcohol, ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-93-2
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00009270
InChI Key LJSQFQKUNVCTIA-UHFFFAOYSA-N
Synonym diethyl sulfide, ethyl sulfide, ethyl thioether, 3-thiapentane, diethylthioether, thioethyl ether, ethylthioethane, 1,1'-thiobisethane, diethyl sulphide, ethane, 1,1'-thiobis
PubChem CID 9609
ChEBI CHEBI:27710
IUPAC Name ethylsulfanylethane
SMILES CCSCC

Specifications

Melting Point -104°C
Density 0.835
Boiling Point 90°C to 92°C
Flash Point −10°C (14°F)
Odor Garlic-like
Refractive Index 1.442
Quantity 100 g
UN Number UN2375
Beilstein 1696909
Merck Index 14,3854
Solubility Information Miscible with alcohol,ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
Formula Weight 90.19
Percent Purity 96%
Chemical Name or Material Diethyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.