missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Sulphate, TRC

Product Code. 30379613
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379613 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379613 Supplier Toronto Research Chemicals Supplier No. TRCD444970250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Sulphate
CAS 64-67-5
Molecular Formula C4H10O4S
Quantity 250 g
Synonym Ethyl sulfate (Et2SO4), DES, Diethyl sulfate, Ethyl sulfate, NSC 56380
InChI Formula InChI=1S/C4H10O4S/c1-3-7-9(5,6)8-4-2/h3-4H2,1-2H3
SMILES CCOS(=O)(=O)OCC
IUPAC Name diethyl sulfate
Molecular Weight (g/mol) 154.18
Formula Weight 154.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.