missing translation for 'onlineSavingsMsg'
Learn More

Diethyl tert-butylmalonate, 96%

Product Code. 11347246
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347246 5 g 5g
11337246 1 g 1g
11357246 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347246 Supplier Thermo Scientific Alfa Aesar Supplier No. L09131.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 759-24-0 | C11H20O4 | 216.28 g/mol

Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl tert-butylmalonate is used in the preparation of enantiomers of 4-tert-butyl-3-isopropyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane 1-sulfide.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 759-24-0
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00009152
InChI Key RJNICNBRGVKNSR-UHFFFAOYSA-N
Synonym diethyl tert-butylmalonate, diethyl t-butylmalonate, diethyl 1,1-dimethylethyl malonate, diethyl 2-tert-butyl malonate, propanedioic acid, 1,1-dimethylethyl-, diethyl ester, diethyltert-butylmalonate, diethyl 2-tert-butylmalonate #, tert-butylmalonic acid diethyl ester, 1,3-diethyl 2-tert-butylpropanedioate
PubChem CID 69798
IUPAC Name diethyl 2-tert-butylpropanedioate
SMILES CCOC(=O)C(C(=O)OCC)C(C)(C)C

Specifications

Density 0.978
Boiling Point 102°C to 104°C (11 mmHg)
Flash Point 90°C (194°F)
Refractive Index 1.425
Quantity 5 g
Beilstein 1781709
Solubility Information Immiscible with water.
Formula Weight 216.28
Percent Purity 97%
Chemical Name or Material Diethyl tert-butylmalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.