missing translation for 'onlineSavingsMsg'
Learn More

Diethyl trans-1,2-cyclopropanedicarboxylate, 97%

Product Code. 10062220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10062220 5 g Glass bottle 5g
10045813 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10062220 Supplier Thermo Scientific Acros Supplier No. 169180050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3999-55-1 | C9H14O4 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3999-55-1
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD00198002
InChI Key SXLDHZFJMXLFJU-RNFRBKRXSA-N
Synonym diethyl trans-1,2-cyclopropanedicarboxylate, trans-diethyl cyclopropane-1,2-dicarboxylate, diethyl trans-cyclopropane-1,2-dicarboxylate, 1r,2r-diethyl cyclopropane-1,2-dicarboxylate, diethyl 1r,2r-cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, diethyl ester, 1r,2r-rel, 1,2-cyclopropanedicarboxylicacid, 1,3-diethyl ester, 1r,2r-rel, 1s,2s-diethyl cyclopropane-1,2-dicarboxylate, diethyltrans-cyclopropane-1,2-dicarboxylate, trans-1,2-diethoxycarbonyl-cyclopropane
PubChem CID 11715303
IUPAC Name diethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
SMILES CCOC(=O)C1CC1C(=O)OCC

Specifications

Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 70°C to 75°C (1.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C3H4(CO2C2H5)2
Refractive Index 1.4400 to 1.442
Quantity 5 g
Specific Gravity 1.06
Formula Weight 186.21
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material Diethyl trans-1, 2-cyclopropanedicarboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.