missing translation for 'onlineSavingsMsg'
Learn More

Diethyl vinylphosphonate, 97%

Product Code. 10465745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10465745 100 g Glass bottle 100g
10410822 10 g Glass bottle 10g
10081711 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10465745 Supplier Thermo Scientific Acros Supplier No. 152151000
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 682-30-4 | C6H13O3P | 164.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 682-30-4
Molecular Formula C6H13O3P
Molecular Weight (g/mol) 164.14
MDL Number MFCD00009079
InChI Key DREPONDJUKIQLX-UHFFFAOYSA-N
Synonym diethyl vinylphosphonate, vinylphosphonic acid diethyl ester, diethyl vinylphosphate, diethoxyphosphinylethene, phosphonic acid, vinyl-, diethyl ester, diethyl ethenylphosphonate, phosphonic acid, ethenyl-, diethyl ester, 1-ethenyl ethoxy phosphoryl oxyethane, acmc-1at5g
PubChem CID 69629
IUPAC Name 1-[ethenyl(ethoxy)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)C=C

Specifications

Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 202°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CH=CH2
Refractive Index 1.4280 to 1.43
Quantity 100 g
Specific Gravity 1.06
Formula Weight 164.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl vinylphosphonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.