missing translation for 'onlineSavingsMsg'
Learn More

Diethylaluminum chloride, 96%, 25% w/w in heptane, Thermo Scientific™

Product Code. 11366389
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11366389 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11366389 Supplier Thermo Scientific Alfa Aesar Supplier No. 089035.ND

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-10-6
Molecular Formula C4H10AlCl
Molecular Weight (g/mol) 120.56
MDL Number MFCD00000459
InChI Key YNLAOSYQHBDIKW-UHFFFAOYSA-M
Synonym diethylaluminum chloride, aluminum, chlorodiethyl, diethylaluminium chloride, deac, chlorodiethylaluminum, diethylchloroaluminum, diethylaluminum monochloride, deak, aluminum diethyl monochloride, aluminum, dichlorotetraethyldi
PubChem CID 7277
SMILES CC[Al](Cl)CC

Specifications

Color Colorless
Flash Point −18°C (−1°F)
Odor Petroleum-like
Linear Formula (C2H5)2AlCl
Quantity 25 g
UN Number UN3394
Sensitivity Air and moisture sensitive
Solubility Information Miscible with hexane.
IUPAC Name chloro(diethyl)alumane
Formula Weight 120.56
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Diethylaluminum chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.