missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, 99+%, extra pure, Thermo Scientific Chemicals

Product Code. 10093073 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal drum
Unit Size:
1 L
10 L
2.5 L
250mL
5mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10093073 250 mL Glass bottle 250mL
10001510 5 mL Glass bottle 5mL
10636922 1 L Glass bottle 1 L
10001500 2.5 L Glass bottle 2.5 L
11975161 10 L Metal drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10093073 Supplier Thermo Scientific Acros Supplier No. 149452500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-89-7
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00009032
InChI Key HPNMFZURTQLUMO-UHFFFAOYSA-N
Synonym diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine
PubChem CID 8021
ChEBI CHEBI:85259
SMILES CCNCC

Specifications

Melting Point -50.0°C
Color Colorless
Density 0.7100g/mL
Boiling Point 55.0°C
Flash Point −23°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2NH
Refractive Index 1.3840 to 1.3860
Quantity 250 mL
Beilstein 04, 95
Fieser 04,148; 13,312; 14,191
Merck Index 15, 3122
Specific Gravity 0.71
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol and most common organic, solvents
IUPAC Name N-ethylethanamine
Viscosity 0.34 mPa.s (25°C)
Formula Weight 73.13
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Diethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.