missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, 99+%, Thermo Scientific Chemicals

Product Code. 11459233
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459233 2500 mL 2500mL
11324499 100 mL 100mL
11449233 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11459233 Supplier Thermo Scientific Alfa Aesar Supplier No. A11716.0F
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Diethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylamine is used as a corrosion inhibitor. It is also used in the production of rubber, resins, dyes and pharmaceuticals. It is also used to cook lysergic acid diethylamide (LSD) and is strictly watched by Drug Enforcement Administration (DEA).

Solubility
Miscible with water, alcohol, ether, chloroform and carbon terachloride.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-89-7
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00009032
InChI Key HPNMFZURTQLUMO-UHFFFAOYSA-N
Synonym diethylamine, n,n-diethylamine, ethanamine, n-ethyl, diethamine, diaethylamin, dwuetyloamina, dietilamina, diethylamin, diethyl amine, n-ethyl-ethanamine
PubChem CID 8021
ChEBI CHEBI:85259
SMILES CCNCC

Specifications

Melting Point °C to 50°C
Density 0.706
Boiling Point 54°C to 55°C
Flash Point -28°C (-18°F)
Assay Percent Range ≥99%
Refractive Index 1.385
Quantity 2500 mL
Beilstein 605268
Sensitivity Air Sensitive
Solubility Information Miscible with water,alcohol,ether,chloroform and carbon terachloride.
IUPAC Name diethylamine
Formula Weight 73.14
Physical Form Liquid
Chemical Name or Material Diethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Diethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.