missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, Extra Pure, SLR, Fisher Chemical™

Product Code. 10408600 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10408600 2.5 L 2.5 L
10253900 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10408600 Supplier Fisher Chemical Supplier No. D/2100/17
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Diethylamine, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-89-7
Molecular Formula C4H11N
MDL Number 9032

Specifications

Melting Point -50°C
Color Colorless
pH 12
Boiling Point 55°C
Packaging Amber glass bottle
Quantity 2.5 L
Formula Weight 73.13g/mol
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diethylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.