missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol, 99%, extra pure

Product Code. 10510561 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10510561 1 L Glass bottle 1 L
10665542 2.5 L Glass bottle 2.5 L
10649083 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10510561 Supplier Thermo Scientific Acros Supplier No. 121160010
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 111-46-6 | C4H10O3 | 106.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-46-6
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00002882
InChI Key MTHSVFCYNBDYFN-UHFFFAOYSA-N
Synonym diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
PubChem CID 8117
ChEBI CHEBI:46807
SMILES OCCOCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -10.0°C
Density 1.1100g/mL
Boiling Point 245.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (HOCH2CH2)2O
Refractive Index 1.4450 to 1.4480
Quantity 1 L
Beilstein 01, 468
Merck Index 15, 3131
Specific Gravity 1.11
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, ether,, ethylene glycol, insoluble in benzene, toluene, carbon, tetrachloride and aliphatic hydrocarbons
IUPAC Name 2-(2-hydroxyethoxy)ethan-1-ol
Viscosity 38 mPa.s (20°C)
Formula Weight 106.12
Percent Purity 99%
Grade Extra Pure
Physical Form Oily Liquid
Chemical Name or Material Diethylene glycol
Show More Show Less
compliance-icons
Product Identifier
  • Diethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.