missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol, 99%

Product Code. 11394489
Change view
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g
Unit Size:
10000g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394489 250 g 250g
11407766 500 g 500g
11417766 2500 g 2500g
11427766 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11394489 Supplier Thermo Scientific Alfa Aesar Supplier No. A14728.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 111-46-6 | C4H10O3 | 106.12 g/mol

Diethylene glycol is used as an important industrial solvent due to its characteristic properties like high boiling point, high viscosity and low vapor loss. It is widely used in the manufacture of unsaturated polyester resins, polyurethanes, nitrocellulose, resins and plasticizers. It plays an important role as a building block in the preparation of morpholine and 1,4-dioxane. It finds application as a heat transfer fluid in the chemical industry due to its low freezing point. As a lubricant, it is used in glass-grinding aids, fiber-finish component and cement-grinding aids. It is also used in natural gas to remove water and other impurities. It is an active component of brake fluid, artificial fog solutions and wallpaper strippers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol is used as an important industrial solvent due to its characteristic properties like high boiling point, high viscosity and low vapor loss. It is widely used in the manufacture of unsaturated polyester resins, polyurethanes, nitrocellulose, resins and plasticizers. It plays an important role as a building block in the preparation of morpholine and 1,4-dioxane. It finds application as a heat transfer fluid in the chemical industry due to its low freezing point. As a lubricant, it is used in glass-grinding aids, fiber-finish component and cement-grinding aids. It is also used in natural gas to remove water and other impurities. It is an active component of brake fluid, artificial fog solutions and wallpaper strippers.

Solubility
Miscible with water, alcohol, ether, acetone, and ethylene glycol.

Notes
Hygroscopic. Heat sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and zinc.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-46-6
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.12
MDL Number MFCD00002882
InChI Key MTHSVFCYNBDYFN-UHFFFAOYSA-N
Synonym diethylene glycol, 2,2'-oxydiethanol, diglycol, diethylenglykol, 2-hydroxyethyl ether, bis 2-hydroxyethyl ether, ethanol, 2,2'-oxybis, 2,2'-oxybisethanol, 2-2-hydroxyethoxy ethanol, digol
PubChem CID 8117
ChEBI CHEBI:46807
SMILES OCCOCCO

Specifications

Melting Point -10°C
Density 1.118
Boiling Point 245°C to 246°C
Flash Point 143°C (289°F)
Assay Percent Range 99%
Odor Nearly Odorless
Linear Formula (HOCH2CH2)2O
Refractive Index 1.447
Quantity 250 g
Beilstein 969209
Sensitivity Hygroscopic
Merck Index 14,3119
Solubility Information Miscible with water,alcohol,ether,acetone,and ethylene glycol.
IUPAC Name 2-(2-hydroxyethoxy)ethan-1-ol
Formula Weight 106.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethylene glycol
Show More Show Less
compliance-icons
Product Identifier
  • Diethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Diethylene glycol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.