missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol diacrylate, tech. 75%, stab.

Product Code. 11352936
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352936 100 g 100g
11342936 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11352936 Supplier Thermo Scientific Alfa Aesar Supplier No. L10682.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4074-88-8 | C10H14O5 | 214.22 g/mol

Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.

Solubility
Slightly soluble in water (1-5g/100ml at 18°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 4074-88-8
Molecular Formula C10H14O5
Molecular Weight (g/mol) 214.22
MDL Number MFCD00014939
InChI Key LEJBBGNFPAFPKQ-UHFFFAOYSA-N
Synonym diethylene glycol diacrylate, oxydiethylene acrylate, oxydiethylene diacrylate, acrylic acid, oxydiethylene ester, tga 2, 2-propenoic acid, oxydi-2,1-ethanediyl ester, diacrylate diethylene glycol, unii-jk01t392ju, acrylic acid, 2-ethoxyethanol diester, di ethylene glycol diacrylate
PubChem CID 19996
IUPAC Name 2-(2-prop-2-enoyloxyethoxy)ethyl prop-2-enoate
SMILES C=CC(=O)OCCOCCOC(=O)C=C

Spécification

Density 1.118
Boiling Point 94°C (0.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.462
Quantity 100 g
UN Number UN2810
Beilstein 1953711
Solubility Information Slightly soluble in water (1-5g/100ml at 18°C).
Formula Weight 214.22
Percent Purity 75%
Grade Technical
Chemical Name or Material Diethylene glycol diacrylate, stabilized
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.