missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol dibutyl ether, 99+%

Product Code. 11346059
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346059 500 mL 500mL
11349807 25 mL 25mL
11339807 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11346059 Supplier Thermo Scientific Alfa Aesar Supplier No. 042150.AP
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extraction of precious metals | CAS: 112-73-2 | C12H26O3 | 218.337 g/mol

Extraction of precious metalsDiethylene glycol dibutyl ether is used as a solvent in Grignard reactions. It is also used as solvents in gold refining, decorative inks for ceramics and digital inks. It finds application in electrochemistry, gas absorption, extractant and high boiling reaction medium. It is also used in fuel, lubricant, textile and medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-73-2
Molecular Formula C12H26O3
Molecular Weight (g/mol) 218.337
MDL Number MFCD00009459
InChI Key KZVBBTZJMSWGTK-UHFFFAOYSA-N
Synonym diethylene glycol dibutyl ether, bis 2-butoxyethyl ether, dibutyl carbitol, butyl diglyme, 1-2-2-butoxyethoxy ethoxy butane, 5,8,11-trioxapentadecane, ether, bis butoxyethyl, 2,2'-dibutoxyethyl ether, ether, bis 2-butoxyethyl
PubChem CID 8210
IUPAC Name 1-[2-(2-butoxyethoxy)ethoxy]butane
SMILES CCCCOCCOCCOCCCC

Specifications

Melting Point -60°C
Color Colorless
Density 0.885 g/mL
Boiling Point 256°C
Flash Point 117°C (243°F)
Refractive Index 1.4235
Quantity 500 mL
Solubility Information Miscible with dimethyl sulfoxide,ethanol and acetone. Immiscible with water.
Formula Weight 218.74
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Diethylene glycol dibutyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.