missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Diethyl Ether, ≥98%, Reagent Grade, Honeywell™

Product Code. 15697840 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Glass bottle
Poly bottle
Stainless steel can
Unit Size:
1 L
20 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15697840 1 L Glass bottle 1 L
15617850 500 mL Glass bottle 500mL
15608490 4 L Poly bottle 4 L
15607850 20 L Stainless steel can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15697840 Supplier Honeywell Research Chemicals Supplier No. E46581L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-36-7
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00009254
InChI Key RRQYJINTUHWNHW-UHFFFAOYSA-N
Synonym diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl
PubChem CID 8179
ChEBI CHEBI:44664
IUPAC Name 1-ethoxy-2-(2-ethoxyethoxy)ethane
SMILES CCOCCOCCOCC

Specifications

Melting Point -44°C
Color Colorless
Density 0.909g/mL at 25°C
Boiling Point 189°C (1013 hPa)
Flash Point 67°C (152.6°F)
Odor Weak
Packaging Glass bottle
Linear Formula (C2H5OCH2CH2)2O
Quantity 1 L
Beilstein 1699259
Solubility Information Soluble in water at 20°C
Vapor Density 5.6 (vs air)
Formula Weight 162.23g/mol
Percent Purity ≥98%
Chemical Name or Material Diethylene glycol diethyl ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.