missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol diethyl ether, 99%

Product Code. 11498077
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11498077

Brand: Thermo Scientific Alfa Aesar A17478.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-36-7 | C8H18O3 | 162.229 g/mol

Diethylene glycol diethyl ether is used as a solvent for reactions performed at higher temperatures. It is involved in the preparation of nitrocellulose, resins and adhesives. It is utilized as a scrubbing medium to absorb carbonyl sulfide (COS), an impurity in petroleum refineries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol diethyl ether is used as a solvent for reactions performed at higher temperatures. It is involved in the preparation of nitrocellulose, resins and adhesives. It is utilized as a scrubbing medium to absorb carbonyl sulfide (COS), an impurity in petroleum refineries.

Solubility
Miscible with hydrocarbons, ethanol, ethyl ether and organic solvents. Immiscible with water.

Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-36-7
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00009254
InChI Key RRQYJINTUHWNHW-UHFFFAOYSA-N
Synonym diethylene glycol diethyl ether, 2-ethoxyethyl ether, diethyl carbitol, ethyl diglyme, 1-ethoxy-2-2-ethoxyethoxy ethane, bis 2-ethoxyethyl ether, diethyldiethylene glycol, 3,6,9-trioxaundecane, degdee, ether, bis 2-ethoxyethyl
PubChem CID 8179
ChEBI CHEBI:44664
IUPAC Name 1-ethoxy-2-(2-ethoxyethoxy)ethane
SMILES CCOCCOCCOCC

Specifications

Melting Point -44°C
Density 0.909
Boiling Point 188°C to 190°C
Flash Point 71°C (160°F)
Odor Weak
Linear Formula (CH3CH2OCH2CH2)2O
Refractive Index 1.412
Quantity 500 mL
Beilstein 1699259
Merck Index 14,3118
Solubility Information Miscible with hydrocarbons,ethanol,ethyl ether and organic solvents. Immiscible with water.
Formula Weight 162.23
Percent Purity 99%
Chemical Name or Material Diethylene glycol diethyl ether
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.