missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Dimethyl Ether, puriss. p.a., ≥99.5% (GC), Honeywell™ Riedel-de-Haën™

Product Code. 15633700
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15633700 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15633700 Supplier Honeywell Riedel-de Haën Supplier No. 322101L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Puriss. p.a.,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Specifications

CAS 111-96-6
Density 0.939g/mL at 25°C, 0.944g/mL at 20°C
Flash Point 57°C (134.6°F)
Packaging Glass bottle
Molecular Formula C6H14O3
Linear Formula (CH3OCH2CH2)2O
MDL Number MFCD00008503
Quantity 1 L
UN Number UN3271
Beilstein 1736101
Synonym diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
InChI Key SBZXBUIDTXKZTM-UHFFFAOYSA-N
SMILES COCCOCCOC
IUPAC Name 1-methoxy-2-(2-methoxyethoxy)ethane
Molecular Weight (g/mol) 134.175
PubChem CID 8150
Vapor Density 4.6 (vs air)
ChEBI CHEBI:46784
Formula Weight 134.17g/mol
Percent Purity ≥99.5% (GC)
Grade puriss. p.a.
Chemical Name or Material Diethylene Glycol Dimethyl Ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.