missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Ethyl Ether, TRC

Product Code. 30135548
Change view
Click to view available options
Quantity:
5 g
250 g
Unit Size:
250g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135548 250 g 250g
17908024 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135548 Supplier Toronto Research Chemicals Supplier No. TRCD444355250G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-90-0
Molecular Formula C6 H14 O3
Molecular Weight (g/mol) 134.17
Synonym Ethanol, 2-(β-ethoxyethoxy)- (3CI), 2-(2-Ethoxyethoxy)ethanol, 1-Hydroxy-3,6-dioxaoctane, 2-(2-Ethoxyethoxy)ethan-1-ol, 2-(2'-Ethoxyethoxy)ethanol, 3,6-Dioxa-1-octanol, Carbitol, Carbitol Solvent Low, Diethylene glycol ethyl ether, Diethylene glycol monoethyl ether, Diglycol monoethyl ether, Dioxitol, Dowanol DE, Ektasolve DE, Ethanol, 2,2'-oxybis-, monoethyl ether, Ethoxydiethylene glycol, Ethyl Diglysolv, Ethyl carbitol, Ethyl digol, Ethyldiethylene glycol, Ethylene diglycol monoethyl ether, NSC 408451, O-Ethyl digol, Poly-Solv DE, Seahosol DG, Seahosol DG-L, Seahosol DG-S, Shihozoru DG, Solvolsol, Transcutol, Transcutol CG, Transcutol HP, Transcutol P, Trivalin SF
IUPAC Name 2-(2-ethoxyethoxy)ethanol
SMILES CCOCCOCCO

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H14O3/c1-2-8-5-6-9-4-3-7/h7H,2-6H2,1H3
Formula Weight 134.0943
Chemical Name or Material Diethylene glycol-monoethylether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.