missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Methyl Ether, ≥99.0%, Reagent grade, Honeywell™

Product Code. 15688430 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass bottle
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15688430 4 L Glass bottle 4 L
15665870 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15688430 Supplier Honeywell Research Chemicals Supplier No. 5795484L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-77-3
Molecular Formula C5H12O3
Molecular Weight (g/mol) 120.15
MDL Number MFCD00002871,MFCD00084416
InChI Key SBASXUCJHJRPEV-UHFFFAOYSA-N
Synonym 2-2-methoxyethoxy ethanol, diethylene glycol monomethyl ether, methyl carbitol, methyl digol, methoxydiglycol, ethanol, 2-2-methoxyethoxy, methyl dioxitol, diethylene glycol methyl ether, dowanol dm, poly-solv dm
PubChem CID 8134
ChEBI CHEBI:44836
IUPAC Name 2-(2-methoxyethoxy)ethan-1-ol
SMILES COCCOCCO

Specifications

Density 1.023g/mL at 25°C
Packaging Glass bottle
Linear Formula CH3OCH2CH2OCH2CH2OH
Quantity 4 L
Beilstein 1697812
Vapor Density 4.14 (vs air)
Formula Weight 120.15g/mol
Percent Purity ≥99.0%
Chemical Name or Material Diethylene glycol methyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.