missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol mono-n-hexyl ether, 97%

Product Code. 11495656
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495656 2500 mL 2500mL
11485656 500 mL 500mL
11405666 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495656 Supplier Thermo Scientific Alfa Aesar Supplier No. B20774.0F
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112-59-4 | C10H22O3 | 190.28 g/mol

Used as a solvent for coatings and degreasing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a solvent for coatings and degreasing.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-59-4
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.28
MDL Number MFCD00010703
InChI Key GZMAAYIALGURDQ-UHFFFAOYSA-N
Synonym diethylene glycol monohexyl ether, 2-2-hexyloxyethoxy ethanol, hexol carbitol, hexyl carbitol, n-hexyl carbitol, n-hexoxyethoxyethanol, 3,6-dioxadodecanol-1, ethanol, 2-2-hexyloxy ethoxy, hexyldiglycol, hexylkarbitol
PubChem CID 8199
IUPAC Name 2-(2-hexoxyethoxy)ethanol
SMILES CCCCCCOCCOCCO

Specifications

Melting Point -40°C
Density 0.932
Boiling Point 259°C to 261°C
Flash Point 123°C (253°F)
Linear Formula HOCH2CH2OCH2CH2O(CH2)5CH3
Refractive Index 1.437
Quantity 2500 mL
Beilstein 1743959
Solubility Information Soluble in water.
Formula Weight 190.28
Percent Purity 97%
Chemical Name or Material Diethylene glycol mono-n-hexyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.