missing translation for 'onlineSavingsMsg'
Learn More

Diethylene glycol monoethyl ether, 98%

Product Code. 11437646
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
This item is not returnable. View return policy

Product Code. 11437646

Brand: Thermo Scientific Alfa Aesar A14670.0C

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 111-90-0 | C6H14O3 | 134.175 g/mol

Diethylene glycol monoethyl ether has low setting point and low viscocity at low temperature so it is used in manufacturing brake fluid. It is used as a flow and gloss promoter in paint industries, in production of printing ink and as a cleaner in offset printing. Also used in textile as a solvent for dyestuff in the printing and dying of fiber & fabrics, in the production & wood preservaties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylene glycol monoethyl ether has low setting point and low viscocity at low temperature so it is used in manufacturing brake fluid. It is used as a flow and gloss promoter in paint industries, in production of printing ink and as a cleaner in offset printing. Also used in textile as a solvent for dyestuff in the printing and dying of fiber & fabrics, in the production & wood preservaties.

Solubility
Fully miscible in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-90-0
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.175
MDL Number MFCD00002872
InChI Key XXJWXESWEXIICW-UHFFFAOYSA-N
Synonym diethylene glycol monoethyl ether, 2-2-ethoxyethoxy ethanol, carbitol, transcutol, ethoxy diglycol, dioxitol, ethyl carbitol, ethyl digol, carbitol solvent, ethanol, 2-2-ethoxyethoxy
PubChem CID 8146
ChEBI CHEBI:40572
IUPAC Name 2-(2-ethoxyethoxy)ethanol
SMILES CCOCCOCCO

Specifications

Melting Point -80°C
Density 0.989
Boiling Point 201°C to 202°C
Flash Point 96°C (204°F)
Odor Pleasant
Linear Formula HOCH2CH2OCH2CH2OCH2CH3
Refractive Index 1.428
Quantity 10,000 g
Beilstein 1736441
Sensitivity Hygroscopic
Merck Index 14,1800
Solubility Information Fully miscible in water.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material Diethylene glycol monoethyl ether
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.