missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriamine, 98+%

Product Code. 11995181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11995181 10 L Plastic drum 10 L
10656682 250 mL Glass bottle 250mL
10090840 1 L Glass bottle 1 L
10510381 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 11995181 Supplier Thermo Scientific Acros Supplier No. 114310100
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Diethylenetriamine, 98+%, C4H13N3, CAS Number-111-40-0, 1,4,7-triazaheptane, 2,2'-diaminodiethylamine, diethylenetriamine, 2,2'-iminodiethylamine, bis 2-aminoethyl amine, barsamide 115, ancamine deta, n,n-bis 2-aminoethyl amine, epicure t, diethylene triamine, 2.5L, 98% min. (GC), 04,255 | CAS: 111-40-0 | C4H13N3 | 103.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-40-0
Molecular Formula C4H13N3
Molecular Weight (g/mol) 103.17
MDL Number MFCD00008171
InChI Key RPNUMPOLZDHAAY-UHFFFAOYSA-N
Synonym diethylenetriamine, bis 2-aminoethyl amine, 2,2'-diaminodiethylamine, diethylene triamine, barsamide 115, epicure t, ancamine deta, 1,4,7-triazaheptane, 2,2'-iminodiethylamine, n,n-bis 2-aminoethyl amine
PubChem CID 8111
ChEBI CHEBI:30629
SMILES NCCNCCN

Specifications

Melting Point -40.0°C
Density 0.9600g/mL
Boiling Point 206.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Plastic drum
Linear Formula H2NCH2CH2NHCH2CH2NH2
Refractive Index 1.4826 to 1.4846
Quantity 10 L
Beilstein 04,255
Specific Gravity 0.96
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most common organic solvents, soluble in hydrocarbons
IUPAC Name bis(2-aminoethyl)amine
Viscosity 5.05 mPa.s (20°C)
Formula Weight 103.17
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Diethylenetriamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.