missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriamine-N,N,N',N″,N″-pentaacetic acid, 90%, MP Biomedicals™

Product Code. 11418111 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418111 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11418111 Supplier MP Biomedicals Supplier No. 0215147891
  • ONLINE EXCLUSIVE PRICE
    240.00 €
    226.80 € / 1kg
    Save 13.20 €
    5% Off
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 67-43-6
Color White
Density 1.770g/mL
Molecular Formula C14H23N3O10
Quantity 1 kg
Synonym pentetic acid, diethylenetriaminepentaacetic acid, dtpa, detapac, complexon v, detarex, perma kleer, titriplex v, monaquest cai, hamp-ex acid
InChI Key QPCDCPDFJACHGM-UHFFFAOYSA-N
SMILES C(CN(CC(=O)O)CC(=O)O)N(CCN(CC(=O)O)CC(=O)O)CC(=O)O
IUPAC Name 2-[bis[2-[bis(carboxymethyl)amino]ethyl]amino]acetic acid
Molecular Weight (g/mol) 393.349
PubChem CID 3053
ChEBI CHEBI:35739
Formula Weight 393.3
Percent Purity 0.9
Physical Form Powder
Chemical Name or Material Diethylenetriamine-N,N,N',N”,N”-pentaacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethylenetriamine-N,N,N',N",N"-pentaacetic Acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Diethylenetriamine-N,N,N',N",N"-pentaacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.