missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Diethylenetriaminepentaacetic acid, pentasodium salt, tech., 40% aqueous solution

Product Code. 10055810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
3 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
3kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10055810 3 kg Glass bottle 3kg
10116613 250 g Glass bottle 250g
10686991 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10055810 Supplier Thermo Scientific Chemicals Supplier No. 407290030
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 140-01-2 | C14H18N3Na5O10 | 503.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylenetriaminepentaacetic acid, pentasodium salt
Name Note 40% Aqueous Solution
CAS 140-01-2, 7732-18-5
Melting Point -40°C
Density 1.2900g/mL
pH 11.0 to 12.0 (1% aq. soln.)
Boiling Point 106°C
Flash Point >100°C
Packaging Glass bottle
Molecular Formula C14H18N3Na5O10
Linear Formula NaOCOCH2N(CH2CH2N(CH2COONa)2)2
Refractive Index 1.4185 to 1.4205
MDL Number MFCD00051016
Quantity 3 kg
Beilstein 04, III, 1190
Specific Gravity 1.29
Synonym pentasodium dtpa, tetralon b, trilon c, versenex 80, pentasodium pentetate, hamp-ex 80, detarex py, kiresuto p, chelest p, plexene d
Solubility Information Solubility in water: soluble. Other solubilities: mixible with many organic solvents
InChI Key LQPLDXQVILYOOL-UHFFFAOYSA-I
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CCN(CC([O-])=O)CC([O-])=O
IUPAC Name pentasodium;2-[bis[2-[bis(carboxylatomethyl)amino]ethyl]amino]acetate
Molecular Weight (g/mol) 503.26
PubChem CID 8779
Formula Weight 503.26
Percent Purity 39 to 41%
Grade Technical
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.