missing translation for 'onlineSavingsMsg'
Learn More

Diethylether, >99%, Stabilized, Multicompendial, Chem-Lab

Product Code. 13298203
Change view
Click to view available options
Quantity:
25 L
Packaging:
MET
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13298203 25 L MET 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13298203 Supplier Chem-Lab Supplier No. CL00.0401.9025
  • ONLINE EXCLUSIVE PRICE
    606.00 €
    358.00 € / 25 L
    Save 248.00 €
    40% Off
 Restricted Item
Estimated Shipment: 03-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pure diethylether stabilized with 5-7ppm BHT for laboratory use. Meets DAB, Ph. Eur., BP specifications.

Specifications

CAS 60-29-7
Density 0.71g/mL
Quantity 25 L
Packaging MET
Molecular Formula C4H10O
MDL Number MFCD00011646
UN Number UN1155
Water <0.2%
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
PubChem CID 3283
ChEBI CHEBI:35702
Formula Weight 74.12g/mol
Percent Purity >99%
Chemical Name or Material Diethylether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethylether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: Diethylether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.